Learn More
Cayman Chemical Methotrexate (hydrate), Purity: ≥98%, CAS: 133073-73-1, 100 mg
SDP
Catalog No. 502954230
Encompass_Preferred
A folic acid and aminopterin derivative; inhibits folic acid metabolism; inhibits purine and pyrimidine synthesis, blocking cell proliferation; induces adenosine release; has anti-inflammatory effects; has been considered the gold standard of DMARD therapy for the immune-inflammatory and joint destructive processes of rheumatoid arthritis
Catalog No. 50-295-4230 Supplier Cayman Chemical Supplier No. 13960100 MG
A folic acid and aminopterin derivative; inhibits folic acid metabolism; inhibits purine and pyrimidine synthesis, blocking cell proliferation; induces adenosine release; has anti-inflammatory effects; has been considered the gold standard of DMARD therapy for the immune-inflammatory and joint destructive processes of rheumatoid arthritis