Learn More
Cayman Chemical Methotrexatehydrate 50mg
SDP
Catalog No. 502955957
Encompass_Preferred
A folic acid and aminopterin derivative; inhibits folic acid metabolism; inhibits purine and pyrimidine synthesis, blocking cell proliferation; induces adenosine release; has anti-inflammatory effects; has been considered the gold standard of DMARD therapy for the immune-inflammatory and joint destructive processes of rheumatoid arthritis
Catalog No. 50-295-5957 Supplier Cayman Chemical Supplier No. 1396050 MG
A folic acid and aminopterin derivative; inhibits folic acid metabolism; inhibits purine and pyrimidine synthesis, blocking cell proliferation; induces adenosine release; has anti-inflammatory effects; has been considered the gold standard of DMARD therapy for the immune-inflammatory and joint destructive processes of rheumatoid arthritis