Learn More
TARGETMOL CHEMICALS INC METHOXAMINE HYDROCHLORID 50MG
SDP
Catalog No. 502581777
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Methoxamine hydrochloride (Methoxamine HCl) is an alpha-1 adrenergic agonist that causes prolonged peripheral VASOCONSTRICTION. purity: 99%
Catalog No. 50-258-1777 Supplier TARGETMOL CHEMICALS INC Supplier No. T099350MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Methoxamine hydrochloride (Methoxamine HCl) is an alpha-1 adrenergic agonist that causes prolonged peripheral VASOCONSTRICTION. purity: 99%