Learn More
Apexbio Technology LLC Methoxamine hydrochloride 61-16-5 10mg
SDP
Catalog No. 5000093283
Encompass_Preferred
Methoxamine hydrochloride (CAS 61-16-5) is a selective agonist of 1-adrenergic receptors leading to vasoconstriction by promoting contraction of vascular smooth muscle This action increases peripheral vascular resistance and raises arterial blood pressure Methoxamine hydrochloride is widely used in biomedical research to model and investigate mechanisms of vasopressor response cardiovascular regulation and adrenergic receptor pharmacology It is also applied experimentally to study hypotensive states particularly those associated with anesthetic or surgical procedures
Catalog No. 50-000-93283 Supplier Apexbio Technology LLC Supplier No. A992210
Methoxamine hydrochloride (CAS 61-16-5) is a selective agonist of 1-adrenergic receptors leading to vasoconstriction by promoting contraction of vascular smooth muscle This action increases peripheral vascular resistance and raises arterial blood pressure Methoxamine hydrochloride is widely used in biomedical research to model and investigate mechanisms of vasopressor response cardiovascular regulation and adrenergic receptor pharmacology It is also applied experimentally to study hypotensive states particularly those associated with anesthetic or surgical procedures