Learn More
Medchemexpress LLC Methoxamine hydrochloride | 61-16-5 | 99.3% | 5 MG
SDP
Catalog No. 5000412021
Encompass_Preferred
Methoxamine hydrochloride is a selective alpha1-adrenergic receptor agonist. It causes vasoconstriction and increased peripheral vascular resistance, and significantly increased the overflow of ATP, ADP, and AMP by a prazosin-sensitive mechanism. This product is for research use only.
  • Selective alpha1-adrenergic receptor agonist.
  • Causes vasoconstriction.
  • Increases peripheral vascular resistance.
  • Increases overflow of ATP, ADP, and AMP.
  • Target: Noradrenergic α1.
  • In vivo studies show greater effect on peripheral resistance vessels than Winkessel vessels in rat systemic circulation.
Catalog No. 50-004-12021 Supplier Medchemexpress LLC Supplier No. HYB12985MG
Methoxamine hydrochloride is a selective alpha1-adrenergic receptor agonist. It causes vasoconstriction and increased peripheral vascular resistance, and significantly increased the overflow of ATP, ADP, and AMP by a prazosin-sensitive mechanism. This product is for research use only.
  • Selective alpha1-adrenergic receptor agonist.
  • Causes vasoconstriction.
  • Increases peripheral vascular resistance.
  • Increases overflow of ATP, ADP, and AMP.
  • Target: Noradrenergic α1.
  • In vivo studies show greater effect on peripheral resistance vessels than Winkessel vessels in rat systemic circulation.