Learn More
Medchemexpress LLC Methoxamine (hydrochloride) | 61-16-5 | 99.3% | 50 MG
SDP
Catalog No. 5000412072
Encompass_Preferred
Methoxamine hydrochloride is a selective alpha1-adrenergic receptor agonist that causes vasoconstriction and increases peripheral vascular resistance. It has been observed to significantly increase the overflow of ATP, ADP, and AMP, but not adenosine, via a prazosin-sensitive mechanism in the rabbit pulmonary artery. It is intended for research use only and is not sold to patients.
  • Selective alpha1-adrenergic receptor agonist
  • Causes vasoconstriction
  • Increases peripheral vascular resistance
  • Increases the overflow of ATP, ADP, and AMP in the rabbit pulmonary artery
Catalog No. 50-004-12072 Supplier Medchemexpress LLC Supplier No. HYB129850MG
Methoxamine hydrochloride is a selective alpha1-adrenergic receptor agonist that causes vasoconstriction and increases peripheral vascular resistance. It has been observed to significantly increase the overflow of ATP, ADP, and AMP, but not adenosine, via a prazosin-sensitive mechanism in the rabbit pulmonary artery. It is intended for research use only and is not sold to patients.
  • Selective alpha1-adrenergic receptor agonist
  • Causes vasoconstriction
  • Increases peripheral vascular resistance
  • Increases the overflow of ATP, ADP, and AMP in the rabbit pulmonary artery