Learn More
Medchemexpress LLC Methoxamine hydrochloride | 61-16-5 | 99.32% | 1 ML
SDP
Catalog No. 5000412020
Encompass_Preferred
Methoxamine hydrochloride is a selective alpha1-adrenergic receptor agonist. It causes vasoconstriction and increased peripheral vascular resistance. It also significantly increased the overflow of ATP, ADP, and AMP, but not adenosine, via a prazosin-sensitive mechanism in the rabbit pulmonary artery.
  • Selective alpha1-adrenergic receptor agonist
  • Causes vasoconstriction
  • Increases peripheral vascular resistance
  • Significantly increases the overflow of ATP, ADP, and AMP (but not adenosine)
  • Acts via a prazosin-sensitive mechanism in the rabbit pulmonary artery
Catalog No. 50-004-12020 Supplier Medchemexpress LLC Supplier No. HYB129810MM/1ML
Methoxamine hydrochloride is a selective alpha1-adrenergic receptor agonist. It causes vasoconstriction and increased peripheral vascular resistance. It also significantly increased the overflow of ATP, ADP, and AMP, but not adenosine, via a prazosin-sensitive mechanism in the rabbit pulmonary artery.
  • Selective alpha1-adrenergic receptor agonist
  • Causes vasoconstriction
  • Increases peripheral vascular resistance
  • Significantly increases the overflow of ATP, ADP, and AMP (but not adenosine)
  • Acts via a prazosin-sensitive mechanism in the rabbit pulmonary artery