Learn More
Selleck Chemical LLC Pyrroloquinoline Quinone Disodium Salt S5263-25mg
SDP
Catalog No. 503112611
Encompass_Preferred
Pyrroloquinoline quinone (methoxatin disodium salt) an aromatic tricyclic o-quinone is a redox cofactor for bacterial dehydrogenases It is an efficient electron transfer catalyst from a number of organic substrates to molecular oxygen (O2) constructing quinoprotein model reactions
Catalog No. 50-311-2611 Supplier Selleck Chemical LLC Supplier No. S526325MG
Pyrroloquinoline quinone (methoxatin disodium salt) an aromatic tricyclic o-quinone is a redox cofactor for bacterial dehydrogenases It is an efficient electron transfer catalyst from a number of organic substrates to molecular oxygen (O2) constructing quinoprotein model reactions