Learn More
Medchemexpress LLC Methoxsalen | 298-81-7 | 99.97% | 216.19 | 50 G
SDP
Catalog No. 5000404323
Encompass_Preferred
Methoxsalen (8-Methoxypsoralen) is a furanocoumarin compound that is utilized in psoralen studies. It is employed in research related to conditions such as psoriasis, eczema, vitiligo, and certain sun-exposed cutaneous lymphomas. Additionally, Methoxsalen acts as a P450 inhibitor. This product is intended for research use only.
  • Furanocoumarin compound.
  • Used in psoralen studies for various skin conditions.
  • Functions as a P450 inhibitor.
  • Available in solid form, white to light yellow in color.
  • High purity.
Catalog No. 50-004-04323 Supplier Medchemexpress LLC Supplier No. HY3015150G
Add to Cart
Add to Cart
Methoxsalen (8-Methoxypsoralen) is a furanocoumarin compound that is utilized in psoralen studies. It is employed in research related to conditions such as psoriasis, eczema, vitiligo, and certain sun-exposed cutaneous lymphomas. Additionally, Methoxsalen acts as a P450 inhibitor. This product is intended for research use only.
  • Furanocoumarin compound.
  • Used in psoralen studies for various skin conditions.
  • Functions as a P450 inhibitor.
  • Available in solid form, white to light yellow in color.
  • High purity.