Learn More
Selleck Chemical LLC Methoxsalen 50mg 298-81-7 Xanthotoxin, NCI-C55903
SDP
Supplier:  Selleck Chemical LLC S195250MG
Encompass_Preferred
Methoxsalen (Xanthotoxin, NCI-C55903) is a naturally occurring photoactive substance found in the seeds of the Ammi majus (Umbelliferae) plant, used in the diagnosis and treatment of psoriasis; A CYP2A5/6 inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-592-0