Learn More
Ace Glass, Inc. 20mL 14/20 methoxy determination boiling flask with capillary side arm.
SDP
Catalog No. 50973242
Encompass
20mL 14/20 methoxy determination boiling flask with capillary side arm.
Catalog No. 50-973-242 Supplier Ace Glass, Inc. Supplier No. 654904
20mL 14/20 methoxy determination boiling flask with capillary side arm.