Learn More
Medchemexpress LLC Methoxy-PEG-C3-amide-C2-Mal | 883993-35-9 | 2000 (average) | (C2H4O)nC11H16N2O4 | 10 MG
SDP
Catalog No. 5000288821
Encompass_Preferred
Methoxy-PEG-C3-amide-C2-Mal is a maleimide-activated methoxy polyethylene glycol (PEG) linker (average MW ~2,000 Da) used to conjugate PEG to thiol-containing biomolecules for site-selective PEGylation in research applications. Supplied as a solid powder and intended for research use only.
  • Maleimide reacts selectively with thiol groups (e.g., cysteine) under mild conditions.
  • Average molecular weight ~2,000 Da for improved solubility and steric shielding.
  • Methoxy end cap minimizes non-specific interactions after conjugation.
  • Supplied as a stable solid; store at recommended cold temperatures to preserve integrity.
  • Available in small lab-scale pack sizes suitable for research workflows.
Catalog No. 50-002-88821 Supplier Medchemexpress LLC Supplier No. HY16089710MG
Methoxy-PEG-C3-amide-C2-Mal is a maleimide-activated methoxy polyethylene glycol (PEG) linker (average MW ~2,000 Da) used to conjugate PEG to thiol-containing biomolecules for site-selective PEGylation in research applications. Supplied as a solid powder and intended for research use only.
  • Maleimide reacts selectively with thiol groups (e.g., cysteine) under mild conditions.
  • Average molecular weight ~2,000 Da for improved solubility and steric shielding.
  • Methoxy end cap minimizes non-specific interactions after conjugation.
  • Supplied as a stable solid; store at recommended cold temperatures to preserve integrity.
  • Available in small lab-scale pack sizes suitable for research workflows.