Learn More
Akina Inc Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Supplier:  Akina Inc AK0291G
Encompass
mPEG-PLGA (Mw ~3,000:36,000 Da) LA:GA 50:50 Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
Catalog No. NC2229288