Learn More
Alamanda Polymers methoxy-poly(ethylene glycol)-block-poly(L-lysine hydrochloride)
SDP
Catalog No. NC1644803
Encompass
mPEG5K-b-PLKC30, average MW 9900, Appearance White to faint-yellow solid, Solubility Clear solution at 50 mg/mL of water
Catalog No. NC1644803 Supplier Alamanda Polymers Supplier No. 050KC030
mPEG5K-b-PLKC30, average MW 9900, Appearance White to faint-yellow solid, Solubility Clear solution at 50 mg/mL of water