Learn More
Akina Inc Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
SDP
Catalog No. NC2827971
Encompass
mPEG-PLGA (Mw ~5000:20000 Da) LA:GA 50:50 Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)
Catalog No. NC2827971 Supplier Akina Inc Supplier No. AK0371G
mPEG-PLGA (Mw ~5000:20000 Da) LA:GA 50:50 Methoxy poly(ethylene glycol)-b-poly(lactide-co-glycolide) (per g)