Learn More
Medchemexpress LLC Methoxy-polyethylene glycol-distearoylphosphatidylethanolamine sodium salt | 95.0% | 3,000 Da (average) | 250 MG
SDP
Catalog No. 5000280742
Encompass_Preferred
m-PEG-DSPE sodium (MW 3000) is a methoxy-polyethylene glycol-conjugated distearoylphosphatidylethanolamine supplied as the sodium salt for research use. It is used as a PEG lipid in nanoparticle and micelle formulations to extend circulation time, improve colloidal stability, and reduce protein adsorption in colloidal drug-delivery systems.
  • PEGylated phospholipid sodium salt for surface modification.
  • Average molecular weight ~3,000 Da reflecting PEG chain length.
  • Purity approximately 95.0% as specified by the manufacturer.
  • Suitable for formulation of liposomes, lipid nanoparticles, and micelles.
  • Improves circulation time and reduces opsonization.
  • Supplied as a small-scale research package for laboratory use.
Catalog No. 50-002-80742 Supplier Medchemexpress LLC Supplier No. HYW440924250MG
m-PEG-DSPE sodium (MW 3000) is a methoxy-polyethylene glycol-conjugated distearoylphosphatidylethanolamine supplied as the sodium salt for research use. It is used as a PEG lipid in nanoparticle and micelle formulations to extend circulation time, improve colloidal stability, and reduce protein adsorption in colloidal drug-delivery systems.
  • PEGylated phospholipid sodium salt for surface modification.
  • Average molecular weight ~3,000 Da reflecting PEG chain length.
  • Purity approximately 95.0% as specified by the manufacturer.
  • Suitable for formulation of liposomes, lipid nanoparticles, and micelles.
  • Improves circulation time and reduces opsonization.
  • Supplied as a small-scale research package for laboratory use.