Learn More
Medchemexpress LLC Methoxy polyethylene glycol N-hydroxysuccinimide ester | 92451-01-9 | 98.0% | 10MG
SDP
Catalog No. 5000260342
Encompass_Preferred
Methoxy polyethylene glycol N-hydroxysuccinimide ester, average molecular weight 20,000 Da, is an NHS-activated methoxy-PEG reagent that reacts efficiently with primary amines to form stable amide bonds. It is supplied as a solid for research use and is commonly used for PEGylation of peptides and proteins, linker construction in targeted degraders, and general bioconjugation to improve solubility and pharmacokinetic properties.
  • Reacts with primary amines to form stable amide linkages.
  • Improves solubility and reduces immunogenicity of conjugates.
  • Provides a 20 kDa hydrophilic spacer for linker design.
  • Supplied as a high-purity reagent suitable for research applications.
  • Compatible with standard coupling and purification workflows.
Catalog No. 50-002-60342 Supplier Medchemexpress LLC Supplier No. HY14070010MG
Methoxy polyethylene glycol N-hydroxysuccinimide ester, average molecular weight 20,000 Da, is an NHS-activated methoxy-PEG reagent that reacts efficiently with primary amines to form stable amide bonds. It is supplied as a solid for research use and is commonly used for PEGylation of peptides and proteins, linker construction in targeted degraders, and general bioconjugation to improve solubility and pharmacokinetic properties.
  • Reacts with primary amines to form stable amide linkages.
  • Improves solubility and reduces immunogenicity of conjugates.
  • Provides a 20 kDa hydrophilic spacer for linker design.
  • Supplied as a high-purity reagent suitable for research applications.
  • Compatible with standard coupling and purification workflows.