Learn More
Medchemexpress LLC Methoxy polyethylene glycol thiol | 134874-49-0 | MFCD00084534 | 90.0% | 750 (average) | (C2H4O)nC3H8OS | 100 MG
SDP
Catalog No. 5000284997
Encompass_Preferred
m-PEG-thiol (MW 750) is a methoxy-terminated polyethylene glycol with a thiol (-SH) end group used to introduce thiol functionality onto biomolecules and surfaces. It facilitates conjugation to gold and other thiol-reactive substrates for nanoparticle functionalization and bioconjugation. The material has an average molecular weight of 750 and a typical purity of 90.0%.
  • Thiol-terminated for efficient conjugation to gold and thiol-reactive chemistries.
  • Average molecular weight 750 for predictable steric and solubility properties.
  • Useful for DNA thiolation and nanoparticle surface modification.
  • Solid form suitable for small-scale reactions and surface treatments.
  • Available in a 100 MG pack for laboratory-scale use.
Catalog No. 50-002-84997 Supplier Medchemexpress LLC Supplier No. HYW591476B100MG
m-PEG-thiol (MW 750) is a methoxy-terminated polyethylene glycol with a thiol (-SH) end group used to introduce thiol functionality onto biomolecules and surfaces. It facilitates conjugation to gold and other thiol-reactive substrates for nanoparticle functionalization and bioconjugation. The material has an average molecular weight of 750 and a typical purity of 90.0%.
  • Thiol-terminated for efficient conjugation to gold and thiol-reactive chemistries.
  • Average molecular weight 750 for predictable steric and solubility properties.
  • Useful for DNA thiolation and nanoparticle surface modification.
  • Solid form suitable for small-scale reactions and surface treatments.
  • Available in a 100 MG pack for laboratory-scale use.