Learn More
TARGETMOL CHEMICALS INC Methoxy-X04 10MG
SDP
Catalog No. 502877060
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Methoxy-X04, a derivative of Congo red and Chrysamine-G, is a brain-permeable fluorescent probe for amyloid-beta (Abeta). Purity 97.01%
Catalog No. 50-287-7060 Supplier TARGETMOL CHEMICALS INC Supplier No. T2297210MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Methoxy-X04, a derivative of Congo red and Chrysamine-G, is a brain-permeable fluorescent probe for amyloid-beta (Abeta). Purity 97.01%