Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methoxy-X04 | 863918-78-9 | MFCD25563254 | >98.0% | 344.40 g/mol | C23H20O3 | 10 MG
SDP

Catalog No. 5000247200
Encompass_Preferred

A brain-penetrant fluorescent probe that selectively binds to β-pleated sheets in amyloid-β plaques and is used for in vivo and ex vivo imaging and quantification of amyloid deposits. Exhibits excitation at 370 nm and emission at 452 nm.

  • Brain-penetrant fluorescent amyloid β probe.
  • Excitation 370 nm, emission 452 nm.
  • Soluble in DMSO (25 mg/mL) and formulatable for in vivo dosing.
  • Recommended storage: protect from light; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-47200 Supplier Medchemexpress LLC Supplier No. HY10324010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

A brain-penetrant fluorescent probe that selectively binds to β-pleated sheets in amyloid-β plaques and is used for in vivo and ex vivo imaging and quantification of amyloid deposits. Exhibits excitation at 370 nm and emission at 452 nm.

  • Brain-penetrant fluorescent amyloid β probe.
  • Excitation 370 nm, emission 452 nm.
  • Soluble in DMSO (25 mg/mL) and formulatable for in vivo dosing.
  • Recommended storage: protect from light; in solvent -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.