Learn More
Abcam Metho x y- x 04, amyloid beta fluorescent marker, 5MG
Catalog No. 502583306
Encompass_Preferred
MW 344.4 Da, Purity >98%. Blood-brain barrier permeable amyloid β fluorescent marker. Blue fluorophore. Derivative of Congo Red (ab145645) and Chrysamine G. Selectively binds fibrillar β-sheet deposits with high affinity (Ki = 26.8 nM). Detects plaques, tangles and cerebrovascular amyloid in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3306 Supplier Abcam Supplier No. AB1428185MG
MW 344.4 Da, Purity >98%. Blood-brain barrier permeable amyloid β fluorescent marker. Blue fluorophore. Derivative of Congo Red (ab145645) and Chrysamine G. Selectively binds fibrillar β-sheet deposits with high affinity (Ki = 26.8 nM). Detects plaques, tangles and cerebrovascular amyloid in vivo. .
The product is subject to the following: Abcam Restricted Use Statement