Learn More
AdipoGen Methoxy2-naphthylacetonitril
SDP
Supplier:  AdipoGen CDXM0072M250
Encompass_Preferred
Chemical. CAS 118736-08-6. Formula C13H11NO. MW 197.23. Synthetic. Fluorescent dye.
Catalog No. 50-256-0545