Learn More
Medchemexpress LLC Methoxyacetic acid | 625-45-6 | MFCD00004308 | 100.0% | 90.08 | C3H6O3 | 1mL
SDP
Catalog No. 5000268057
Encompass_Preferred
Methoxyacetic acid is a metabolite of ethylene glycol monomethyl ether When the concentration of methoxyacetic acid reaches a certain level it can inhibit the respiratory function of hepatic mitochondria and testicular mitochondria Methoxyacetic acid is somewhat toxic[1][2]
Catalog No. 50-002-68057 Supplier Medchemexpress LLC Supplier No. HYY100910MM/1ML
Add to Cart
Add to Cart
Methoxyacetic acid is a metabolite of ethylene glycol monomethyl ether When the concentration of methoxyacetic acid reaches a certain level it can inhibit the respiratory function of hepatic mitochondria and testicular mitochondria Methoxyacetic acid is somewhat toxic[1][2]