Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methoxyacetyl chloride

Catalog No. M965310G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M965310G 10 g
M96531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M965310G Supplier Sigma Aldrich Supplier No. M965310G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methoxyacetyl chloride can be used for methoxyacetylation of:
  • Bilocularin a to synthesize 8-methoxyacetoxybilocularin a
  • Substituted and unsubstituted benzamides to synthesize n-(2-methoxyacetyl)-benzamides.it can also be used for the synthesis of α-lactams by reacting with imines. and in the post-synthetic modification of the metal-organic framework (mof), mil-101, to synthesize a catalyst for cycloaddition reaction of co2 with propylene oxide

Specifications

CAS 38870-89-2
Density 1.187 g/mL (at 25°C (literature))
Boiling Point 112°C to 113°C (lit.)
Molecular Formula C3H5ClO2
Refractive Index n20/D 1.419 (literature)
Linear Formula CH3OCH2COCl
MDL Number MFCD00000728
Quantity 10 g
Synonym 2-methoxyacetyl chloride; Methoxyacetic acid chloride
Molecular Weight (g/mol) 108.52
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.