Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetyl chloride, 97%, stabilized

Catalog No. AC125700100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125700100 10 g Glass bottle
AC125700500 50 g Glass bottle
AC125702500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC125700100 Supplier Thermo Scientific Chemicals Supplier No. 125700100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38870-89-2 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38870-89-2
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000728
InChI Key JJKWHOSQTYYFAE-UHFFFAOYSA-N
Synonym methoxyacetyl chloride, methoxyacetylchloride, acetyl chloride, methoxy, methoxy-acetyl chloride, methoxyacetic acid chloride, unii-t66s02b1s3, methoxy acetylchloride, 2-methoxyacetylchloride, methoxy acetyl chloride, ch3och2cocl
PubChem CID 96623
IUPAC Name 2-methoxyacetyl chloride
SMILES COCC(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.1870g/mL
Boiling Point 112°C to 113°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4190 to 1.422
Linear Formula CH3OCH2COCl
Quantity 10 g
Fieser 12,310
Specific Gravity 1.187
Solubility Information Solubility in water: reacts
Stabilizer 0.3%mgO
Formula Weight 108.52
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methoxyacetyl chloride, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 254-169-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.