Learn More
Medchemexpress LLC Methoxyamine-d3 hydrochloride | 110220-55-8 | 99.5% | 86.54 g·mol^-1 | CH3D3ClNO | 100mg
SDP
Catalog No. 5000268150
Encompass_Preferred
Methoxyamine-d3 (O-Methylhydroxylamine-d3) hydrochloride is the deuterium labeled Methoxyamine hydrochloride Methoxyamine hydrochloride is an orally active and potent base excision repair (BER) inhibitor[1]
Catalog No. 50-002-68150 Supplier Medchemexpress LLC Supplier No. HYY0958S100MG
Methoxyamine-d3 (O-Methylhydroxylamine-d3) hydrochloride is the deuterium labeled Methoxyamine hydrochloride Methoxyamine hydrochloride is an orally active and potent base excision repair (BER) inhibitor[1]