Learn More
Medchemexpress LLC Methoxychlor-d6 | 106031-79-2 | 98.2% | 1 MG
SDP
Catalog No. 5000410819
Encompass_Preferred
Methoxychlor-d6 is a deuterium-labeled Methoxychlor, designed for research use. It serves as a tracer and internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-004-10819 Supplier Medchemexpress LLC Supplier No. HYB1873S1MG
Methoxychlor-d6 is a deuterium-labeled Methoxychlor, designed for research use. It serves as a tracer and internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated into drug molecules for quantitation during drug development
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs through deuteration