Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxychlor 95.0+%, TCI America™
SDP

Catalog No. M173625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M173625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M173625G Supplier TCI America Supplier No. M173625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 72-43-5
Molecular Formula C16H15Cl3O2
Molecular Weight (g/mol) 345.644
MDL Number MFCD00000803
InChI Key IAKOZHOLGAGEJT-UHFFFAOYSA-N
Synonym methoxychlor, p,p'-methoxychlor, dmdt, dimethoxy-ddt, methoxy-ddt, methoxcide, metox, maralate, marlate, moxie
PubChem CID 4115
ChEBI CHEBI:6842
IUPAC Name 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl

Specifications

Melting Point 87°C
Quantity 25 g
UN Number 3077
Formula Weight 345.64
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methoxychlor
RTECSNumber KJ3675000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.