Learn More
Cayman Chemical Methoxyfenozide, Purity: ≥98%, CAS: 161050-58-4, 100 mg
SDP
Catalog No. 502939910
Encompass_Preferred
A diacylhydrazine insecticide; selectively binds to lepidopteran EcRs over dipteran EcRs (Kds = 0.5 and 124 nM, respectively); lethal to neonatal larvae of S. exigua, S. frugiperda, T. ni, O. nubilalis, L. pomonella, H. zea, and H. virescens (LC50s = 0.35, 0.2, 0.11, 0.18, 0.21, 0.79, and 3.12 mg/L, respectively); induces early molting in O. nubilalis neonatal larvae at 0.063-0.5 ppm
Catalog No. 50-293-9910 Supplier Cayman Chemical Supplier No. 29182100 MG
A diacylhydrazine insecticide; selectively binds to lepidopteran EcRs over dipteran EcRs (Kds = 0.5 and 124 nM, respectively); lethal to neonatal larvae of S. exigua, S. frugiperda, T. ni, O. nubilalis, L. pomonella, H. zea, and H. virescens (LC50s = 0.35, 0.2, 0.11, 0.18, 0.21, 0.79, and 3.12 mg/L, respectively); induces early molting in O. nubilalis neonatal larvae at 0.063-0.5 ppm