Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methoxyfenozide-d9 | 2469014-53-5 | 99.0% | 377.52 | C22H19D9N2O3 | 5 MG
SDP

Catalog No. 5000228812
Encompass_Preferred

Methoxyfenozide-d9 is a deuterium-labeled analog of methoxyfenozide used as a stable isotope internal standard for quantitative analytical methods. It is intended for use as a tracer or internal standard in techniques such as NMR, GC-MS, and LC-MS, offering high purity and defined isotopic enrichment for reliable quantitation.

  • Deuterium-labeled internal standard for analytical methods (NMR, GC-MS, LC-MS).
  • High purity: 99%.
  • Chemical formula: C22H19D9N2O3.
  • Molecular weight: 377.52.
  • Available as a 5 mg powder.
  • Store powder at -20°C; in solvent store at -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-28812 Supplier Medchemexpress LLC Supplier No. HY117386S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methoxyfenozide-d9 is a deuterium-labeled analog of methoxyfenozide used as a stable isotope internal standard for quantitative analytical methods. It is intended for use as a tracer or internal standard in techniques such as NMR, GC-MS, and LC-MS, offering high purity and defined isotopic enrichment for reliable quantitation.

  • Deuterium-labeled internal standard for analytical methods (NMR, GC-MS, LC-MS).
  • High purity: 99%.
  • Chemical formula: C22H19D9N2O3.
  • Molecular weight: 377.52.
  • Available as a 5 mg powder.
  • Store powder at -20°C; in solvent store at -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.