Learn More
Medchemexpress LLC Methoxyfenozide-d9 | 2469014-53-5 | 99.0% | 377.52 g/mol | C22H19D9N2O3 | 10 MG
SDP
Catalog No. 5000227591
Encompass_Preferred
Methoxyfenozide-d9 is a deuterium-labeled analytical standard of methoxyfenozide supplied as a white to off-white solid. It is intended for use as an internal standard in quantitative analysis and method development where isotopically labeled analogs improve measurement accuracy and precision.
  • Deuterium-labeled (d9) analog for isotope-dilution methods.
  • High purity suitable for analytical use.
  • High isotopic enrichment for accurate mass discrimination.
  • Stable solid form with recommended cold storage for longevity.
  • Available in a 10 mg package for small-scale analytical workflows.
Catalog No. 50-002-27591 Supplier Medchemexpress LLC Supplier No. HY117386S10MG
Methoxyfenozide-d9 is a deuterium-labeled analytical standard of methoxyfenozide supplied as a white to off-white solid. It is intended for use as an internal standard in quantitative analysis and method development where isotopically labeled analogs improve measurement accuracy and precision.
  • Deuterium-labeled (d9) analog for isotope-dilution methods.
  • High purity suitable for analytical use.
  • High isotopic enrichment for accurate mass discrimination.
  • Stable solid form with recommended cold storage for longevity.
  • Available in a 10 mg package for small-scale analytical workflows.