Learn More
Nanocs Methoxyl PEG, 1g
Supplier:  Nanocs MPEG40K
Encompass
Molecular weight: 160~40000 Da. Appearance: Viscous liquid or white solid depends on MW. Moisture Content: <0.5%. Molecular wieght: +-15% around its mean MW. Nanocs’ Methoxypolyethylene glycols (MPEGs) are water-soluble linear polymers formed by the ringopening addition of ethylene oxide units to methoxy end unit with different molecular weight ranges from 160 to 40 kD. These products are non-toxic, non-irritating, have good solubility in water, does not hydrolyze and deteriorate, with very narrow molecular weight distribution. Their properties make them among the most versatile chemical ingredients and processing aids available to formulators and manufacturers of a wide range of products.
Catalog No. NC1362791