Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methoxymethyl propionate | 70767-92-9 | 98.0% | 118.13 | 1 G
SDP

Catalog No. 5000404582
Encompass_Preferred

Methoxymethyl propionate is a biological molecule intended for research use only. It has not been fully validated for medical applications and is not sold to patients. This product is for experienced personnel.

  • Colorless to light yellow liquid appearance
  • Molecular formula of C₅H₁₀O₃
  • Storage in pure form at -20°C for 3 years or 4°C for 2 years
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL (846.53 mM)
  • In Vivo dissolution methods available for clear solutions

Catalog No. 50-004-04582 Supplier Medchemexpress LLC Supplier No. HY416161G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methoxymethyl propionate is a biological molecule intended for research use only. It has not been fully validated for medical applications and is not sold to patients. This product is for experienced personnel.

  • Colorless to light yellow liquid appearance
  • Molecular formula of C₅H₁₀O₃
  • Storage in pure form at -20°C for 3 years or 4°C for 2 years
  • Storage in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL (846.53 mM)
  • In Vivo dissolution methods available for clear solutions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.