Learn More
Medchemexpress LLC Methoxymethyl propionate | 70767-92-9 | 98.0% | 118.13 | 250 MG
SDP
Catalog No. 5000402424
Encompass_Preferred
Methoxymethyl propionate is a biological molecule intended for research use only and is not sold to patients. This chemical is provided in liquid form, typically colorless to light yellow, and offers high purity for various laboratory applications.
  • High purity at 98.0% for reliable results.
  • Available in liquid form, colorless to light yellow.
  • Suitable for research applications.
  • Soluble in DMSO with specific preparation guidelines.
  • Stable for extended periods under recommended storage conditions: -20°C for 3 years (pure form) and -80°C for 6 months (in solvent).
Catalog No. 50-004-02424 Supplier Medchemexpress LLC Supplier No. 250MGHY41616
Methoxymethyl propionate is a biological molecule intended for research use only and is not sold to patients. This chemical is provided in liquid form, typically colorless to light yellow, and offers high purity for various laboratory applications.
  • High purity at 98.0% for reliable results.
  • Available in liquid form, colorless to light yellow.
  • Suitable for research applications.
  • Soluble in DMSO with specific preparation guidelines.
  • Stable for extended periods under recommended storage conditions: -20°C for 3 years (pure form) and -80°C for 6 months (in solvent).