Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methoxymethyl propionate | 70767-92-9 | 98.0% | 118.13 | 50 MG
SDP

Catalog No. 5000404530
Encompass_Preferred

Methoxymethyl propionate is a biological molecule intended for research use only. MedChemExpress does not sell this product to patients.

  • Purity of 98.0%
  • Available in various quantities from 50 mg to 5 g (larger quantities available via quote)
  • Ships at room temperature in continental US
  • Storage recommendations: Pure form at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)
  • Provides in vivo dissolution protocols using various co-solvents

Catalog No. 50-004-04530 Supplier Medchemexpress LLC Supplier No. HY4161650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methoxymethyl propionate is a biological molecule intended for research use only. MedChemExpress does not sell this product to patients.

  • Purity of 98.0%
  • Available in various quantities from 50 mg to 5 g (larger quantities available via quote)
  • Ships at room temperature in continental US
  • Storage recommendations: Pure form at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (100 mg/mL)
  • Provides in vivo dissolution protocols using various co-solvents

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.