Learn More
Medchemexpress LLC Methoxymethyl propionate | 70767-92-9 | 98.0% | 118.13 | 500 MG
SDP
Catalog No. 5000404529
Encompass_Preferred
Methoxymethyl propionate is a biological molecule appearing as a colorless to light yellow liquid, intended for research use only. It is not for sale to patients. For in vitro use, it can be dissolved in DMSO at a concentration of 100 mg/mL. In vivo experiments offer various dissolution protocols.
  • Suitable for research use only
  • Appears as a colorless to light yellow liquid
  • Soluble in DMSO for in vitro applications
  • Offers multiple dissolution protocols for in vivo experiments
  • Should be stored at room temperature for shipping
  • Pure form storage recommended at -20°C for 3 years or 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-04529 Supplier Medchemexpress LLC Supplier No. HY41616500MG
Methoxymethyl propionate is a biological molecule appearing as a colorless to light yellow liquid, intended for research use only. It is not for sale to patients. For in vitro use, it can be dissolved in DMSO at a concentration of 100 mg/mL. In vivo experiments offer various dissolution protocols.
  • Suitable for research use only
  • Appears as a colorless to light yellow liquid
  • Soluble in DMSO for in vitro applications
  • Offers multiple dissolution protocols for in vivo experiments
  • Should be stored at room temperature for shipping
  • Pure form storage recommended at -20°C for 3 years or 4°C for 2 years
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month