Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyphenamine Hydrochloride, MP Biomedicals™

Catalog No. ICN15542925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15542925 25 g
ICN15542901 1 g
ICN15542905 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15542925 Supplier MP Biomedicals, Inc Supplier No. 0215542925

missing translation for 'validMainframeMsg'

Soluble in water (50mg/mL)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-3-5588
Molecular Formula C11H18ClNO
Molecular Weight (g/mol) 215.72
MDL Number MFCD00034832
InChI Key WJVCROKJTIUCAZ-UHFFFAOYNA-N

Specifications

Color White
Quantity 25 g
Synonym methoxyphenamine hydrochloride, methoxyphenamine hcl, orthoxine hydrochloride, ortodrinex hydrochloride, methoxyphenaminium chloride, methoxiphenadrin hydrochloride, o-methoxymethamphetamine hydrochloride, 1-2-methoxyphenyl propan-2-yl methyl amine hydrochloride, 1-2-methoxyphenyl-n-methylpropan-2-amine hydrochloride
SMILES CC(CC1=CC=CC=C1OC)NC.Cl
IUPAC Name 1-(2-methoxyphenyl)-N-methylpropan-2-amine;hydrochloride
PubChem CID 9859211
Formula Weight 215.7
Physical Form Powder
Chemical Name or Material Methoxyphenamine Hydrochloride
EINECSNumber 226-993-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.