Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methoxypolyethylene Glycol Amine

Catalog No. 066791G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-679-1G 1 g
06-679-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-679-1G Supplier Sigma Aldrich Supplier No. 066791G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methoxypolyethylene glycol amine is generally used for the chemical modification of biologically active compounds and therapeutic agents. It has been used to increase the aqueous solubility of long-wavelength voltage-sensitive dyes and to prepare the PEG-coated fluorescent beads in studying large-scale rotational cytoplasmic flow.

Specifications

CAS 80506-64-5
Linear Formula H2NCH2CH2(OCH2CH2)nOCH3
MDL Number MFCD00131599
Quantity 1 g
Synonym O-(2-Aminoethyl)-O'-methylpolyethylene glycol; Aminopolyethylene glycol monomethyl ether; Methoxypolyoxyethylene amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.