Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methoxypolyethylene Glycol Amine

Catalog No. 07964250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-964-250MG 250 mg
07-964-1G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-964-250MG Supplier Sigma Aldrich Supplier No. 07964250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methoxypolyethylene glycol amine can:Form dendritic micelles to enhance the solubility of hydrophobic compounds.Conjugate with an anticancer drug methotrexate (MTX) to form MTX-PEG with potential application as drug carriers.Couple with polyethylenimine (PEI) to for mgraft copolymers which can be used as nonviral gene vectors.Oligomer used to prepare PEG-ylated conjugates having the advantage of reduced immunogenicity and increased serum half life

Specifications

CAS 80506-64-5
Linear Formula H2NCH2CH2(OCH2CH2)nOCH3
MDL Number MFCD00131599
Quantity 250 mg
Synonym mPEG-NH2; O-(2-Aminoethyl)-O'-methylpolyethylene glycol; Aminopolyethylene glycol monomethyl ether; Methoxypolyoxyethylene amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.