Learn More
Medchemexpress LLC Methoxypolyethylene glycol maleimide | 99126-64-4 | 99.9% | 5000 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY166990500MG
Encompass_Preferred
Methoxypolyethylene glycol maleimide (MW 5000) (MeO-PEG-Mal) is a protein PEGylation reagent. For research use only.
  • Protein PEGylation reagent
  • Suitable for research applications
Catalog No. 50-003-91155