Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methoxypolyethylene glycol maleimide (MW 5000) | 99126-64-4 | 99.9% | 5000 | 250 MG
SDP

Catalog No. 5000392580
Encompass_Preferred

Methoxypolyethylene glycol maleimide (MW 5000) is a protein PEGylation reagent.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: -20°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Catalog No. 50-003-92580 Supplier Medchemexpress LLC Supplier No. 250MGHY166990
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methoxypolyethylene glycol maleimide (MW 5000) is a protein PEGylation reagent.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: -20°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.