Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methoxytrimethylsilane

Catalog No. 25300625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-300-625G 25 g
25-300-6250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-300-625G Supplier Sigma Aldrich Supplier No. 25300625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methoxytrimethylsilane (MTMS) is a silylating agent, which is majorly used to provide a hydrophobic surface coating with a high water contact angle and low surface energy.Superhydrophobic recycled polyethylene terephthalate (rPET) aerogels can be fabricated by using rPET fibers and polyvinyl alcohol (PVA) and can be coated on MTMS. MTMS on diglycidyl bisphenol A resin (epoxy resin) also shows weathering resistance on exposure to UV by lowering the effect of color change by 45%.

Specifications

CAS 1825-61-2
Density 0.756 g/mL (at 25°C)
Boiling Point 57°C to 58°C
Molecular Formula C4H12OSi
Refractive Index n20/D 1.366 (literature)
Linear Formula CH3OSi(CH3)3
Quantity 25 g
MDL Number MFCD00008344
Synonym Methyl trimethylsilyl ether; Trimethylmethoxysilane
Molecular Weight (g/mol) 104.22
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.