Learn More
Apexbio Technology LLC Methscopolamine 155-41-9 200mg
SDP
Catalog No. 5000090626
Encompass_Preferred
Methscopolamine is a small-molecule antagonist targeting muscarinic acetylcholine receptors It is designed to inhibit these receptors thereby reducing acetylcholine-mediated signaling and regulating gastrointestinal secretions and motility Methscopolamine exerts its biological activity primarily through antagonism of muscarinic cholinergic receptors Based on these pharmacological properties methscopolamine holds research potential in studying muscarinic receptor-mediated physiological processes particularly gastrointestinal functions and secretory mechanisms and in experimental models evaluating peripheral cholinergic receptor contributions to gastrointestinal disorders
Catalog No. 50-000-90626 Supplier Apexbio Technology LLC Supplier No. B1611200
Methscopolamine is a small-molecule antagonist targeting muscarinic acetylcholine receptors It is designed to inhibit these receptors thereby reducing acetylcholine-mediated signaling and regulating gastrointestinal secretions and motility Methscopolamine exerts its biological activity primarily through antagonism of muscarinic cholinergic receptors Based on these pharmacological properties methscopolamine holds research potential in studying muscarinic receptor-mediated physiological processes particularly gastrointestinal functions and secretory mechanisms and in experimental models evaluating peripheral cholinergic receptor contributions to gastrointestinal disorders