Learn More
TARGETMOL CHEMICALS INC Methscopolamine 200MG
SDP
Catalog No. 502880500
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Methscopolamine (Hyoscine methyl bromide) (Pamine), as a natural plant alkaloid, is a muscarinic acetylcholine receptor blocker. Purity 99.7%
Catalog No. 50-288-0500 Supplier TARGETMOL CHEMICALS INC Supplier No. T0253200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Methscopolamine (Hyoscine methyl bromide) (Pamine), as a natural plant alkaloid, is a muscarinic acetylcholine receptor blocker. Purity 99.7%