Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methscopolamine bromide | 155-41-9 | 99.8% | 1 ML
SDP

Catalog No. 5000406208
Encompass_Preferred

Methscopolamine Bromide is a biochemical reagent suitable for life science research. It is designed for various biological applications and is for research use only.

  • Molecular Weight: 398.29
  • Formula: C18H24BrNO4
  • Appearance: White to off-white solid
  • Soluble in DMSO and H2O
  • Store solid at 4°C, sealed and away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Catalog No. 50-004-06208 Supplier Medchemexpress LLC Supplier No. HYB034410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methscopolamine Bromide is a biochemical reagent suitable for life science research. It is designed for various biological applications and is for research use only.

  • Molecular Weight: 398.29
  • Formula: C18H24BrNO4
  • Appearance: White to off-white solid
  • Soluble in DMSO and H2O
  • Store solid at 4°C, sealed and away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.