Learn More
Medchemexpress LLC Methscopolamine bromide | 155-41-9 | 99.81% | 500 MG
SDP
Catalog No. 5000406278
Encompass_Preferred
Methscopolamine bromide is a biochemical reagent utilized in life science research. It functions as a biological material or organic compound, specifically for research applications, and is not intended for patient use.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Available in solid, white to off-white form
  • Soluble in DMSO and H2O
  • Documentation such as data sheet, COA, and SDS available
  • Related to GPCR/G Protein and neuronal signaling
  • Acts as a muscarinic acetylcholine receptor inhibitor
Catalog No. 50-004-06278 Supplier Medchemexpress LLC Supplier No. HYB0344500MG
Methscopolamine bromide is a biochemical reagent utilized in life science research. It functions as a biological material or organic compound, specifically for research applications, and is not intended for patient use.
  • Biochemical reagent for life science research
  • Functions as a biological material or organic compound
  • Available in solid, white to off-white form
  • Soluble in DMSO and H2O
  • Documentation such as data sheet, COA, and SDS available
  • Related to GPCR/G Protein and neuronal signaling
  • Acts as a muscarinic acetylcholine receptor inhibitor