Learn More
Cayman Chemical Methsuximide, Purity: ≥98%, CAS: 77-41-8, 100 mg
SDP
Catalog No. 502981179
Encompass_Preferred
An anticonvulsant; inhibits CYP2C19-mediated metabolism of chlorproguanil and proguanil at 100 µM; protects against leptazol-induced convulsions in rats when administered at a dose of 100 mg/kg
Catalog No. 50-298-1179 Supplier Cayman Chemical Supplier No. 19058100 MG
An anticonvulsant; inhibits CYP2C19-mediated metabolism of chlorproguanil and proguanil at 100 µM; protects against leptazol-induced convulsions in rats when administered at a dose of 100 mg/kg