Learn More
Cayman Chemical MethsuxImIde 10mg
SDP
Catalog No. 502983629
Encompass_Preferred
An anticonvulsant; inhibits CYP2C19-mediated metabolism of chlorproguanil and proguanil at 100 µM; protects against leptazol-induced convulsions in rats when administered at a dose of 100 mg/kg
Catalog No. 50-298-3629 Supplier Cayman Chemical Supplier No. 1905810 MG
An anticonvulsant; inhibits CYP2C19-mediated metabolism of chlorproguanil and proguanil at 100 µM; protects against leptazol-induced convulsions in rats when administered at a dose of 100 mg/kg