Learn More
Cayman Chemical Methyclothiazide, Purity: ≥98%, CAS: 135-07-9, 10 mg
SDP
Catalog No. 502994843
Encompass_Preferred
A thiazide diuretic; inhibits norepinephrine- and vasopressin-induced contractions of aortic rings isolated from spontaneously hypertensive rats at a concentration of 100 μM; reduces salt-induced increases in blood pressure in salt-sensitive but not salt-resistant rats without affecting body weight or heart rate
Catalog No. 50-299-4843 Supplier Cayman Chemical Supplier No. 2379910 MG
A thiazide diuretic; inhibits norepinephrine- and vasopressin-induced contractions of aortic rings isolated from spontaneously hypertensive rats at a concentration of 100 μM; reduces salt-induced increases in blood pressure in salt-sensitive but not salt-resistant rats without affecting body weight or heart rate