Learn More
Medchemexpress LLC Methyclothiazide | 135-07-9 | MFCD00242610 | 99.8% | 360.24 | C9H11Cl2N3O4S2 | 10 MG
SDP
Catalog No. 5000298998
Encompass_Preferred
Methyclothiazide is a thiazide-class diuretic and antihypertensive research reagent used in pharmacology and in vitro studies to investigate diuretic mechanisms, vascular responsiveness, and voltage-dependent calcium channel activity.
  • Thiazide-class diuretic used to study diuresis and blood pressure regulation.
  • Reduces vascular response to vasoconstrictors and modulates voltage-dependent calcium channel activity in vitro.
  • High measured purity suitable for analytical and pharmacological research.
  • Supplied as a small-quantity solid suitable for laboratory assays and method development.
  • Accompanied by a datasheet containing CAS, formula, and molecular weight for reproducible experiments.
Catalog No. 50-002-98998 Supplier Medchemexpress LLC Supplier No. HYB056210MG
Methyclothiazide is a thiazide-class diuretic and antihypertensive research reagent used in pharmacology and in vitro studies to investigate diuretic mechanisms, vascular responsiveness, and voltage-dependent calcium channel activity.
  • Thiazide-class diuretic used to study diuresis and blood pressure regulation.
  • Reduces vascular response to vasoconstrictors and modulates voltage-dependent calcium channel activity in vitro.
  • High measured purity suitable for analytical and pharmacological research.
  • Supplied as a small-quantity solid suitable for laboratory assays and method development.
  • Accompanied by a datasheet containing CAS, formula, and molecular weight for reproducible experiments.